siRNA Details
| Gene Symbol: GRAMD1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172105.1 | NP_001165576.1 | 6 | 937 | ||
| NM_017577.4 | NP_060047.3 | 11 | 1210 | |||
| XM_005247546.2 | XP_005247603.1 | 10 | 1126 | |||
| XM_005247547.1 | XP_005247604.1 | 11 | 1172 | |||
| XM_011512930.1 | XP_011511232.1 | 11 | 1205 | |||
| XM_011512931.1 | XP_011511233.1 | 6 | 994 | |||
| XM_017006646.1 | XP_016862135.1 | 10 | 1124 | |||
| XM_017006647.1 | XP_016862136.1 | 12 | 1288 | |||
| XM_017006648.1 | XP_016862137.1 | 7 | 1017 | |||
| XM_017006649.1 | XP_016862138.1 | 5 | 768 | |||
| GenBank mRNA | AK093181.1 | - | 6 | 729 | ||
| AK097783.1 | - | 12 | 1294 | |||
| AK292846.1 | - | 11 | 1146 | |||
| AK292964.1 | - | 6 | 852 | |||
| AK293884.1 | - | 9 | 922 | |||
| AK304142.1 | - | 6 | 937 | |||
| AL133661.1 | - | 2 | 1426 | |||
| AY358725.1 | - | 6 | 890 | |||
| BC028972.2 | - | 3 | 214 | |||
| BC035040.1 | - | 11 | 1216 | 4829130 | ||

GRAMD1C
Genomic Map