siRNA Details
| Gene Symbol: KLHL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001257194.1 | NP_001244123.1 | 4 | 969 | ||
| NM_001257195.1 | NP_001244124.1 | 2 | 545 | |||
| NM_017415.2 | NP_059111.2 | 4 | 794 | |||
| GenBank mRNA | AB032955.1 | - | 4 | 564 | ||
| AF208068.1 | - | 4 | 478 | |||
| AF208069.1 | - | 2 | 582 | |||
| AF208070.1 | - | 4 | 969 | |||
| AK303916.1 | - | 2 | 545 | |||
| AK310568.1 | - | 2 | 282 | |||
| AK314707.1 | - | 4 | 535 | |||
| BC034035.1 | - | 4 | 793 | 4817650 | ||

KLHL3
Genomic Map