siRNA Details
| Gene Symbol: ZRANB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017580.2 | NP_060050.2 | 1 | 560 | ||
| XM_005269925.4 | XP_005269982.1 | 2 | 415 | |||
| XM_005269926.4 | XP_005269983.1 | 2 | 603 | |||
| XM_006717907.2 | XP_006717970.1 | 2 | 1285 | |||
| XM_017016357.1 | XP_016871846.1 | 2 | 2016 | |||
| XM_017016358.1 | XP_016871847.1 | 1 | 128 | |||
| GenBank mRNA | AJ252060.1 | - | 1 | 595 | ||
| AK302810.1 | - | 2 | 371 | |||
| AK307386.1 | - | 1 | 200 | |||

ZRANB1
Genomic Map