siRNA Details
| Gene Symbol: PLXNA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017514.4 | NP_059984.3 | 31, 32 | 5552 | ||
| XM_005274705.4 | XP_005274762.2 | 31, 32 | 5492 | |||
| XM_005274706.4 | XP_005274763.2 | 31, 32 | 5224 | |||
| XM_006724829.3 | XP_006724892.1 | 31, 32 | 5456 | |||
| XM_011531183.2 | XP_011529485.1 | 30, 31 | 4916 | |||
| GenBank mRNA | X87852.1 | - | 31, 32 | 5561 | ||

PLXNA3
Genomic Map