siRNA Details
Gene Symbol: HYDIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198542.1 | NP_001185471.1 | 8 | 973 | ||
NM_001198543.1 | NP_001185472.1 | 8 | 943 | |||
NM_001270974.2 | NP_001257903.1 | 8 | 1041 | |||
NM_017558.4 | NP_060028.2 | 8 | 1041 | |||
XM_006721206.2 | XP_006721269.1 | 8 | 935 | |||
XM_011523146.2 | XP_011521448.1 | 8 | 968 | |||
XM_011523147.1 | XP_011521449.1 | 8 | 935 | |||
XM_011523148.1 | XP_011521450.1 | 8 | 1008 | |||
XM_011523151.1 | XP_011521453.1 | 8 | 967 | |||
XM_017023346.1 | XP_016878835.1 | 8 | 1213 | |||
GenBank mRNA | AK022933.1 | - | 8 | 975 | ||
AK299016.1 | - | 8 | 930 | |||
AK299348.1 | - | 8 | 973 | |||
AK308296.1 | - | 6 | 560 | |||
BC028351.1 | - | 8 | 922 |
Gene Symbol: HYDIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103556.1 | 3 | 221 | |||
GenBank mRNA | AL833826.1 | - | 3 | 221 |
