siRNA Details
| Gene Symbol: KIF21B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252100.1 | NP_001239029.1 | 35 | 8853 | ||
| NM_001252102.1 | NP_001239031.1 | 35 | 8237 | |||
| NM_001252103.1 | NP_001239032.1 | 34 | 8198 | |||
| NM_017596.3 | NP_060066.2 | 34 | 8814 | |||
| GenBank mRNA | AB007918.2 | - | 34 | 8814 | ||
| AL137395.1 | - | 1 | 2581 | |||
| BC052964.1 | - | 1 | 2266 | |||
| BC078676.1 | - | 6 | 3821 | |||

KIF21B
Genomic Map