siRNA Details
| Gene Symbol: KIF18B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001264573.1 | NP_001251503.1 | 15 | 3137 | ||
| NM_001265577.1 | NP_001252506.1 | 16 | 2874 | |||
| XM_011524385.2 | XP_011522687.1 | 16 | 3211 | |||
| XM_011524386.2 | XP_011522688.1 | 16 | 2945 | |||
| XM_011524387.2 | XP_011522689.1 | 16 | 3185 | |||
| XM_011524388.2 | XP_011522690.1 | 16 | 3175 | |||
| XM_011524389.2 | XP_011522691.1 | 15 | 3438 | |||
| XM_011524390.2 | XP_011522692.1 | 15 | 3107 | |||
| GenBank mRNA | BC044933.1 | - | 6 | 1770 | ||
| BC067365.1 | - | 3 | 1299 | |||

KIF18B
Genomic Map