siRNA Details
| Gene Symbol: IFIT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270927.1 | NP_001257856.1 | 3 | 1280 | ||
| NM_001270928.1 | NP_001257857.1 | 3 | 1297 | |||
| NM_001270929.1 | NP_001257858.1 | 3 | 1292 | |||
| NM_001270930.1 | NP_001257859.1 | 3 | 1418 | |||
| NM_001548.4 | NP_001539.3 | 2 | 1183 | |||
| GenBank mRNA | AK092813.1 | - | 2 | 2211 | ||
| AK308688.1 | - | 3 | 1184 | |||
| AK314588.1 | - | 2 | 1106 | |||
| BC007091.1 | - | 2 | 1108 | 4250452 | ||
| BT006667.1 | - | 1 | 1016 | |||
| X03557.1 | - | 2 | 1080 | |||

IFIT1
Genomic Map