siRNA Details
| Gene Symbol: ZNF264 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003417.4 | NP_003408.1 | 4 | 3235 | ||
| XM_011527522.2 | XP_011525824.1 | 3 | 3139 | |||
| GenBank mRNA | AB007872.1 | - | 4 | 3184 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF264| Gene Symbol: ZNF264 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003417.4 | NP_003408.1 | 4 | 3235 | ||
| XM_011527522.2 | XP_011525824.1 | 3 | 3139 | |||
| GenBank mRNA | AB007872.1 | - | 4 | 3184 | ||