siRNA Details
Gene Symbol: Amy1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010970.1 | NP_001010970.1 | 4 | 454 | ||
GenBank mRNA | AB057450.1 | - | 4 | 454 | ||
BC088228.1 | - | 3 | 397 | 7323054 | ||
FQ219213.1 | - | 4 | 783 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
