siRNA Details
Gene Symbol: Prps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017243.2 | NP_058939.1 | 7 | 1311 | ||
GenBank mRNA | CO405720.1 | - | 2 | 461 | ||
X16554.1 | - | 7 | 1308 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Prps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017243.2 | NP_058939.1 | 7 | 1311 | ||
GenBank mRNA | CO405720.1 | - | 2 | 461 | ||
X16554.1 | - | 7 | 1308 |