siRNA Details
| Gene Symbol: Arhgef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021694.3 | NP_067726.3 | 28 | 2927 | ||
| GenBank mRNA | BC091218.1 | - | 28 | 2927 | 7107802 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arhgef1| Gene Symbol: Arhgef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021694.3 | NP_067726.3 | 28 | 2927 | ||
| GenBank mRNA | BC091218.1 | - | 28 | 2927 | 7107802 | |