siRNA Details
| Gene Symbol: Cgnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK173252.1 | - | 17 | 4043 | ||
| BC026522.1 | - | 1 | 358 | |||
| BC031499.1 | - | 3 | 1653 | |||
| BC038477.1 | - | 1 | 212 | |||
| BC039211.1 | - | 11 | 2760 | |||
| BC108380.1 | - | 1 | 360 | |||
| BC139006.1 | - | 18 | 5179 | 8862028 | ||
| BC158061.1 | - | 18 | 5189 | 9007361 | ||

Cgnl1
Genomic Map