siRNA Details
| Gene Symbol: Cgnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB186125.1 | - | 2 | 829 | ||
| AK036871.1 | - | 2 | 394 | |||
| AK040774.1 | - | 2 | 350 | |||
| AK081179.1 | - | 2 | 353 | |||
| BB629895.1 | - | 2 | 394 | |||
| BC139006.1 | - | 1 | 304 | 8862028 | ||
| BC158061.1 | - | 1 | 304 | 9007361 | ||

Cgnl1
Genomic Map