siRNA Details
| Gene Symbol: Rtn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009953.2 | NP_001009953.2 | 7 | 919 | ||
| NM_080909.2 | NP_543185.2 | 8 | 3093 | |||
| XM_006230649.2 | XP_006230711.1 | 9 | 3165 | |||
| XM_006230650.2 | XP_006230712.1 | 7 | 889 | |||
| GenBank mRNA | AF442357.1 | - | 7 | 922 | ||
| AF442358.1 | - | 7 | 922 | |||
| AY164698.1 | - | 7 | 922 | |||
| AY496443.1 | - | 8 | 3093 | |||
| BC062068.1 | - | 7 | 917 | |||
| BC107448.1 | - | 7 | 919 | |||
| BC129077.1 | - | 7 | 928 | |||

Rtn3
Genomic Map