siRNA Details
| Gene Symbol: Opa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133585.3 | NP_598269.3 | 18, 19 | 1899 | ||
| XM_006248497.1 | XP_006248559.1 | 20, 21 | 2163 | |||
| XM_006248498.1 | XP_006248560.1 | 19, 20 | 2109 | |||
| XM_006248499.1 | XP_006248561.1 | 19, 20 | 2052 | |||
| GenBank mRNA | AY333988.1 | - | 18, 19 | 1808 | ||
| AY510274.1 | - | 18, 19 | 1954 | |||
| BC111071.1 | - | 18, 19 | 1899 | 7324017 | ||
| U93197.1 | - | 2, 3 | 105 | |||

Opa1
Genomic Map