siRNA Details
| Gene Symbol: Opa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133585.3 | NP_598269.3 | 24, 25 | 2630 | ||
| XM_006248497.1 | XP_006248559.1 | 26, 27 | 2894 | |||
| XM_006248498.1 | XP_006248560.1 | 25, 26 | 2840 | |||
| XM_006248499.1 | XP_006248561.1 | 25, 26 | 2783 | |||
| GenBank mRNA | AY333988.1 | - | 24, 25 | 2539 | ||
| AY510274.1 | - | 24, 25 | 2685 | |||
| BC111071.1 | - | 24, 25 | 2630 | 7324017 | ||
| U93197.1 | - | 8, 9 | 836 | |||

Opa1
Genomic Map