siRNA Details
| Gene Symbol: Cav1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031556.3 | NP_113744.2 | 3 | 597 | ||
| NM_133651.2 | NP_598412.2 | 2 | 589 | |||
| XM_006236130.2 | XP_006236192.1 | 3 | 569 | |||
| XM_006236131.1 | XP_006236193.1 | 3 | 536 | |||
| GenBank mRNA | AF439778.1 | - | 3 | 442 | ||
| AF439779.1 | - | 2 | 349 | |||
| BC078744.1 | - | 3 | 476 | |||
| BC104689.1 | - | 2 | 412 | |||
| BC128717.1 | - | 2 | 419 | |||
| BC161826.1 | - | 2 | 582 | 9100114 | ||
| Z46614.1 | - | 3 | 470 | |||

Cav1
Genomic Map