siRNA Details
| Gene Symbol: CARMIL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138360.3 | NP_612369.3 | 10 | 867 | ||
| XM_011537300.2 | XP_011535602.1 | 9 | 526 | |||
| XM_017021751.1 | XP_016877240.1 | 6 | 427 | |||
| GenBank mRNA | BG913168.1 | - | 1 | 19 | ||
| BG977591.1 | - | 1 | 80 | |||
| DT932621.1 | - | 1 | 37 | |||

CARMIL3
Genomic Map