siRNA Details
| Gene Symbol: Tmem87a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110496.1 | NP_001103966.1 | 1 | 136 | ||
| NM_001110497.1 | NP_001103967.1 | 1 | 136 | |||
| NM_173734.3 | NP_776095.2 | 1 | 136 | |||
| GenBank mRNA | AK044591.1 | - | 1 | 90 | ||
| AK086738.1 | - | 1 | 86 | |||
| AK088559.1 | - | 1 | 118 | |||
| AK134268.1 | - | 1 | 99 | |||
| AK137137.1 | - | 1 | 102 | |||
| BC004047.1 | - | 1 | 85 | |||
| BC027354.1 | - | 1 | 109 | 4010477 | ||

Tmem87a
Genomic Map