siRNA Details
| Gene Symbol: Tmem87a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110496.1 | NP_001103966.1 | 20 | 2601 | ||
| NM_001110497.1 | NP_001103967.1 | 20 | 2598 | |||
| NM_173734.3 | NP_776095.2 | 20 | 2604 | |||
| GenBank mRNA | AK044591.1 | - | 20 | 2555 | ||
| AK088559.1 | - | 19 | 2509 | |||
| AK168093.1 | - | 1 | 934 | |||
| BC004047.1 | - | 20 | 2550 | |||
| BC024676.1 | - | 10 | 1552 | |||
| BC027354.1 | - | 20 | 2577 | 4010477 | ||

Tmem87a
Genomic Map