siRNA Details
| Gene Symbol: Tmem87a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110496.1 | NP_001103966.1 | 20 | 2825 | ||
| NM_001110497.1 | NP_001103967.1 | 20 | 2822 | |||
| NM_173734.3 | NP_776095.2 | 20 | 2828 | |||
| GenBank mRNA | AK044591.1 | - | 21 | 2757 | ||
| AK088559.1 | - | 20 | 2711 | |||
| AK168093.1 | - | 2 | 1134 | |||
| BC004047.1 | - | 21 | 2751 | |||
| BC024676.1 | - | 11 | 1753 | |||
| BC027354.1 | - | 21 | 2779 | 4010477 | ||

Tmem87a
Genomic Map