siRNA Details
| Gene Symbol: Fnip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173753.4 | NP_776114.2 | 14 | 1778 | ||
| GenBank mRNA | AK087849.1 | - | 14 | 2432 | ||
| AK122574.1 | - | 14 | 1726 | |||
| BC079892.1 | - | 14 | 1768 | 30543296 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fnip1| Gene Symbol: Fnip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173753.4 | NP_776114.2 | 14 | 1778 | ||
| GenBank mRNA | AK087849.1 | - | 14 | 2432 | ||
| AK122574.1 | - | 14 | 1726 | |||
| BC079892.1 | - | 14 | 1768 | 30543296 | ||