siRNA Details
| Gene Symbol: Ctsr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175581.3 | NP_783171.2 | 6 | 851 | ||
| XM_008771434.1 | XP_008769656.1 | 5 | 772 | |||
| GenBank mRNA | AF456459.1 | - | 811 | |||
| BC097484.1 | - | 6 | 851 | 7459242 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctsr| Gene Symbol: Ctsr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175581.3 | NP_783171.2 | 6 | 851 | ||
| XM_008771434.1 | XP_008769656.1 | 5 | 772 | |||
| GenBank mRNA | AF456459.1 | - | 811 | |||
| BC097484.1 | - | 6 | 851 | 7459242 | ||