siRNA Details
| Gene Symbol: Trappc11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177240.3 | NP_796214.2 | 6, 7 | 839 | ||
| GenBank mRNA | AK083445.1 | - | 6, 7 | 836 | ||
| AK160812.1 | - | 6, 7 | 840 | |||
| BC063065.1 | - | 1, 2 | 52 | |||
| BC147504.1 | - | 6, 7 | 757 | 9056419 | ||
| BC151159.1 | - | 6, 7 | 757 | 9088048 | ||

Trappc11
Genomic Map