siRNA Details
| Gene Symbol: Trappc11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177240.3 | NP_796214.2 | 23, 24 | 2810 | ||
| GenBank mRNA | AK083445.1 | - | 23, 24 | 2808 | ||
| AK160812.1 | - | 22, 23 | 3134 | |||
| BC063065.1 | - | 16, 17 | 2420 | |||
| BC094246.1 | - | 7, 8 | 892 | |||
| BC147504.1 | - | 23, 24 | 2728 | 9056419 | ||
| BC151159.1 | - | 23, 24 | 2728 | 9088048 | ||

Trappc11
Genomic Map