siRNA Details
| Gene Symbol: Fastkd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177244.3 | NP_796218.2 | 7 | 1650 | ||
| GenBank mRNA | AK220423.1 | - | 3 | 585 | ||
| BC034292.1 | - | 4 | 757 | |||
| BC069943.1 | - | 6 | 1143 | |||
| BC075689.1 | - | 7 | 1650 | 30609755 | ||

Fastkd1
Genomic Map