siRNA Details
| Gene Symbol: Fastkd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177244.3 | NP_796218.2 | 12 | 2351 | ||
| GenBank mRNA | AK220423.1 | - | 8 | 1199 | ||
| BC023501.1 | - | 5 | 484 | |||
| BC034292.1 | - | 9 | 1458 | |||
| BC069943.1 | - | 11 | 1844 | |||
| BC075689.1 | - | 12 | 2351 | 30609755 | ||

Fastkd1
Genomic Map