siRNA Details
| Gene Symbol: Clec16a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204229.1 | NP_001191158.1 | 23 | 5983 | ||
| NM_177562.5 | NP_808230.2 | 24 | 5989 | |||
| GenBank mRNA | AK028051.1 | - | 1 | 2243 | ||
| AK046244.1 | - | 1 | 1095 | |||
| AK122255.1 | - | 24 | 5941 | |||
| BC049280.1 | - | 1 | 1404 | |||

Clec16a
Genomic Map