siRNA Details
| Gene Symbol: TMEM26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178505.7 | NP_848600.2 | 6 | 4473 | ||
| GenBank mRNA | CR749606.1 | - | 6 | 4473 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM26| Gene Symbol: TMEM26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178505.7 | NP_848600.2 | 6 | 4473 | ||
| GenBank mRNA | CR749606.1 | - | 6 | 4473 | ||