siRNA Details
| Gene Symbol: CPLX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181654.3 | NP_857637.1 | 3 | 1285 | ||
| GenBank mRNA | AB593158.1 | - | 3 | 1266 | ||
| AK123184.1 | - | 3 | 1285 | |||
| BX640655.1 | - | 3 | 1264 | |||
| BX648518.1 | - | 3 | 1268 | |||

CPLX4
Genomic Map