siRNA Details
| Gene Symbol: C22orf15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK093880.1 | - | 3 | 747 | ||
| BC113450.1 | - | 1 | 109 | 8322502 | ||
| BC113452.1 | - | 1 | 109 | 8322504 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C22orf15| Gene Symbol: C22orf15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK093880.1 | - | 3 | 747 | ||
| BC113450.1 | - | 1 | 109 | 8322502 | ||
| BC113452.1 | - | 1 | 109 | 8322504 | ||