siRNA Details
| Gene Symbol: Fam53b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175268.4 | NP_780477.2 | 4 | 520 | ||
| NM_212473.1 | NP_997638.1 | 4 | 583 | |||
| GenBank mRNA | AK020830.1 | - | 4 | 521 | ||
| AK031576.1 | - | 3 | 364 | |||
| AK051299.1 | - | 4 | 584 | |||
| AK147350.1 | - | 4 | 916 | |||
| AK147725.1 | - | 4 | 712 | |||
| AK156629.1 | - | 4 | 483 | |||
| AK195672.1 | - | 3 | 287 | |||
| BC035523.1 | - | 4 | 515 | 5400742 | ||

Fam53b
Genomic Map