siRNA Details
| Gene Symbol: Fam53b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175268.4 | NP_780477.2 | 5 | 1963 | ||
| NM_212473.1 | NP_997638.1 | 5 | 2026 | |||
| GenBank mRNA | AK031576.1 | - | 4 | 1807 | ||
| AK051299.1 | - | 5 | 2027 | |||
| AK147350.1 | - | 5 | 2359 | |||
| AK147725.1 | - | 5 | 2156 | |||
| AK165022.1 | - | 1241 | ||||
| AK220076.1 | - | 1 | 129 | |||
| BC035523.1 | - | 5 | 1958 | 5400742 | ||

Fam53b
Genomic Map