siRNA Details
| Gene Symbol: Habp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001505.1 | NP_001001505.1 | 13 | 1783 | ||
| XM_006231633.2 | XP_006231695.1 | 12 | 1717 | |||
| GenBank mRNA | AB177406.1 | - | 13 | 1783 | ||
| BC129080.1 | - | 12 | 1669 | 8374257 | ||

Habp2
Genomic Map