siRNA Details
| Gene Symbol: Lppr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001508.2 | NP_001001508.2 | 7 | 1990 | ||
| GenBank mRNA | AF541280.1 | - | 7 | 1991 | ||
| AY266268.1 | - | 7 | 1991 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lppr4| Gene Symbol: Lppr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001508.2 | NP_001001508.2 | 7 | 1990 | ||
| GenBank mRNA | AF541280.1 | - | 7 | 1991 | ||
| AY266268.1 | - | 7 | 1991 | |||