siRNA Details
| Gene Symbol: Slc25a47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001509.1 | NP_001001509.1 | 6 | 1362 | ||
| GenBank mRNA | AY603424.1 | - | 6 | 1362 | ||
| BC089874.1 | - | 6 | 1366 | 7325453 | ||
| FQ210765.1 | - | 6 | 1422 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc25a47| Gene Symbol: Slc25a47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001509.1 | NP_001001509.1 | 6 | 1362 | ||
| GenBank mRNA | AY603424.1 | - | 6 | 1362 | ||
| BC089874.1 | - | 6 | 1366 | 7325453 | ||
| FQ210765.1 | - | 6 | 1422 | |||