siRNA Details
| Gene Symbol: Usp19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001516.2 | NP_001001516.1 | 20 | 2947 | ||
| XM_006243793.2 | XP_006243855.1 | 21 | 3265 | |||
| XM_006243794.2 | XP_006243856.1 | 21 | 3262 | |||
| XM_006243795.2 | XP_006243857.1 | 21 | 3259 | |||
| XM_006243796.2 | XP_006243858.1 | 21 | 3241 | |||
| XM_006243797.2 | XP_006243859.1 | 21 | 3264 | |||
| XM_006243798.2 | XP_006243860.1 | 21 | 3261 | |||
| XM_006243799.2 | XP_006243861.1 | 21 | 3258 | |||
| GenBank mRNA | AY605065.1 | - | 20 | 2947 | ||
| BC168243.1 | - | 20 | 3273 | |||

Usp19
Genomic Map