siRNA Details
| Gene Symbol: GIPC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304725.1 | NP_001291654.1 | 2 | 196 | ||
| NM_017655.5 | NP_060125.4 | 2 | 493 | |||
| GenBank mRNA | AB073737.1 | - | 2 | 309 | ||
| AK000082.1 | - | 1 | 52 | |||
| AK225941.1 | - | 2 | 332 | |||
| AK307422.1 | - | 2 | 196 | |||
| BC036075.1 | - | 2 | 493 | 5296057 | ||
| BI561167.1 | - | 2 | 499 | |||

GIPC2
Genomic Map