siRNA Details
| Gene Symbol: rnf141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001800.1 | NP_001001800.1 | 4 | 436 | ||
| XM_006230050.2 | XP_006230112.1 | 4 | 456 | |||
| XM_008759730.1 | XP_008757952.1 | 5 | 562 | |||
| XM_008759731.1 | XP_008757953.1 | 4 | 429 | |||
| GenBank mRNA | AY621087.1 | - | 4 | 436 | ||

rnf141
Genomic Map