siRNA Details
| Gene Symbol: Nol4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001986.2 | NP_001001986.2 | 8 | 4148 | ||
| NM_001134300.2 | NP_001127772.1 | 11 | 4933 | |||
| GenBank mRNA | AK018446.1 | - | 1 | 1516 | ||
| AK147524.1 | - | 11 | 4936 | |||
| AK164107.1 | - | 1 | 2553 | |||
| AK164526.1 | - | 3 | 2939 | |||
| BC029060.1 | - | 1 | 2910 | |||
| BC075648.1 | - | 10 | 4343 | 30620653 | ||
| BC079552.1 | - | 8 | 4090 | |||
| BC094234.1 | - | 7 | 3999 | |||

Nol4l
Genomic Map