siRNA Details
| Gene Symbol: GPATCH8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002909.3 | NP_001002909.1 | 8 | 892 | ||
| NM_001304939.1 | NP_001291868.1 | 7 | 817 | |||
| NM_001304940.1 | NP_001291869.1 | 10 | 1033 | |||
| NM_001304941.1 | NP_001291870.1 | 9 | 910 | |||
| NM_001304942.1 | NP_001291871.1 | 7 | 819 | |||
| NM_001304943.1 | NP_001291872.1 | 5 | 773 | |||
| XM_011524557.1 | XP_011522859.1 | 10 | 966 | |||
| XM_011524558.1 | XP_011522860.1 | 11 | 785 | |||
| XM_011524559.2 | XP_011522861.1 | 12 | 863 | |||
| XM_011524561.2 | XP_011522863.1 | 8 | 624 | |||
| XM_017024372.1 | XP_016879861.1 | 13 | 908 | |||
| XM_017024373.1 | XP_016879862.1 | 10 | 1743 | |||
| XM_017024374.1 | XP_016879863.1 | 12 | 826 | |||
| XM_017024375.1 | XP_016879864.1 | 9 | 921 | |||
| XM_017024376.1 | XP_016879865.1 | 10 | 781 | |||
| XM_017024377.1 | XP_016879866.1 | 9 | 758 | |||
| XM_017024378.1 | XP_016879867.1 | 10 | 939 | |||
| XM_017024379.1 | XP_016879868.1 | 11 | 821 | |||
| XM_017024380.1 | XP_016879869.1 | 10 | 759 | |||
| GenBank mRNA | AB011125.2 | - | 7 | 1982 | ||
| AK025600.1 | - | 8 | 694 | |||
| BC136629.1 | - | 9 | 709 | 9020619 | ||
| BC144323.1 | - | 9 | 709 | |||
| CB161089.1 | - | 4 | 345 | |||
| CN388517.1 | - | 7 | 597 | |||
| CN388519.1 | - | 7 | 608 | |||

GPATCH8
Genomic Map