siRNA Details
Gene Symbol: CCDC144NL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004306.2 | NP_001004306.1 | 4 | 2602 | ||
GenBank mRNA | BC068597.1 | - | 4 | 2602 | 4837813 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CCDC144NL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004306.2 | NP_001004306.1 | 4 | 2602 | ||
GenBank mRNA | BC068597.1 | - | 4 | 2602 | 4837813 |