siRNA Details
Gene Symbol: ZNF774 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004309.2 | NP_001004309.2 | 4 | 1819 | ||
XM_017022127.1 | XP_016877616.1 | 5 | 1827 | |||
XM_017022128.1 | XP_016877617.1 | 3 | 2501 | |||
XM_017022129.1 | XP_016877618.1 | 4 | 1691 | |||
GenBank mRNA | BC067279.1 | - | 4 | 1754 | 6181677 |
