siRNA Details
Gene Symbol: ZNF705A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004328.2 | NP_001004328.1 | 5 | 2860 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZNF705A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004328.2 | NP_001004328.1 | 5 | 2860 |