siRNA Details
| Gene Symbol: BAIAP2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026857.1 | 2 | 2219 | |||
| GenBank mRNA | AK027350.1 | - | 1 | 83 | ||
| AK056555.1 | - | 2 | 2201 | |||
| AK075238.1 | - | 1 | 182 | |||
| AK096609.1 | - | 1 | 1127 | |||
| AK124762.1 | - | 2 | 2260 | |||
| BC110822.1 | - | 1 | 228 | |||

BAIAP2-AS1
Genomic Map