siRNA Details
| Gene Symbol: ZYG11A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004339.2 | NP_001004339.2 | 11 | 2245 | ||
| NM_001307931.1 | NP_001294860.1 | 10 | 1493 | |||
| XM_011541480.2 | XP_011539782.1 | 10 | 1975 | |||
| XM_011541481.2 | XP_011539783.1 | 10 | 1843 | |||
| XM_011541484.1 | XP_011539786.1 | 11 | 2020 | |||
| GenBank mRNA | AY271826.1 | - | 10 | 1484 | ||

ZYG11A
Genomic Map