siRNA Details
| Gene Symbol: RNF213 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256071.2 | NP_001243000.2 | 29 | 7745 | ||
| XM_005257545.4 | XP_005257602.2 | 30 | 7933 | |||
| XM_005257546.4 | XP_005257603.2 | 30 | 7837 | |||
| XM_006721995.3 | XP_006722058.1 | 30 | 8220 | |||
| XM_011525084.2 | XP_011523386.1 | 30 | 7933 | |||
| XM_011525086.2 | XP_011523388.1 | 30 | 7933 | |||
| XM_011525087.2 | XP_011523389.1 | 30 | 7933 | |||
| XM_017024905.1 | XP_016880394.1 | 25 | 6800 | |||
| GenBank mRNA | AB537889.1 | - | 29 | 7738 | ||

RNF213
Genomic Map