siRNA Details
| Gene Symbol: Lrp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031322.3 | NP_112612.2 | 34 | 5051 | ||
| XM_008762038.1 | XP_008760260.1 | 34 | 5263 | |||
| GenBank mRNA | AB011533.1 | - | 21 | 3217 | ||
| AB073317.3 | - | 34 | 5051 | |||

Lrp4
Genomic Map