siRNA Details
| Gene Symbol: FERMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017671.4 | NP_060141.3 | 2 | 918 | ||
| GenBank mRNA | AB105105.1 | - | 2 | 918 | ||
| AF443278.1 | - | 2 | 487 | |||
| AK314394.1 | - | 2 | 308 | |||
| AY137240.1 | - | 1 | 129 | |||
| BC035882.1 | - | 2 | 601 | 4941567 | ||

FERMT1
Genomic Map